Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145288.1 | NP_660331.1 | 3 | 597 | ||
GenBank mRNA | AF447583.1 | - | 4 | 643 | ||
AY040679.1 | - | 2 | 193 | |||
BC019352.1 | - | 3 | 597 | 3942704 |