Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077624.1 | NP_001071092.1 | 6 | 920 | ||
XM_005259772.3 | XP_005259829.1 | 6 | 711 | |||
XM_005259773.4 | XP_005259830.1 | 6 | 917 | |||
XM_005259775.3 | XP_005259832.1 | 6 | 708 | |||
XM_006722658.3 | XP_006722721.1 | 6 | 920 | |||
XM_011527717.2 | XP_011526019.1 | 6 | 723 | |||
XM_011527718.2 | XP_011526020.1 | 6 | 720 | |||
XM_011527719.2 | XP_011526021.1 | 6 | 739 | |||
XM_011527720.2 | XP_011526022.1 | 5 | 494 | |||
XM_011527721.2 | XP_011526023.1 | 4 | 268 | |||
XM_017026404.1 | XP_016881893.1 | 6 | 878 | |||
XM_017026405.1 | XP_016881894.1 | 5 | 1360 | |||
XM_017026406.1 | XP_016881895.1 | 3 | 198 | |||
XM_017026407.1 | XP_016881896.1 | 6 | 920 | |||
XM_017026408.1 | XP_016881897.1 | 6 | 611 | |||
GenBank mRNA | AK097652.1 | - | 5 | 1013 | ||
AK289536.1 | - | 2 | 521 | |||
AK308938.1 | - | 5 | 720 | |||
BC118576.1 | - | 6 | 491 | 40053684 | ||
BC118657.1 | - | 5 | 408 |