Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 13, 14 | 1258 | ||
NM_001300858.1 | NP_001287787.1 | 13, 14 | 1409 | |||
NM_144977.4 | NP_659414.2 | 13, 14 | 1258 | |||
NR_125340.1 | 12, 13 | 1198 | ||||
XM_005244931.2 | XP_005244988.1 | 12, 13 | 1198 | |||
XM_006711192.2 | XP_006711255.1 | 14, 15 | 1168 | |||
XM_006711193.3 | XP_006711256.1 | 14, 15 | 1300 | |||
XM_011509246.2 | XP_011507548.1 | 13, 14 | 1119 | |||
XM_011509247.1 | XP_011507549.1 | 12, 13 | 1059 | |||
XM_011509248.2 | XP_011507550.1 | 13, 14 | 1052 | |||
XM_011509249.2 | XP_011507551.1 | 14, 15 | 1357 | |||
XM_011509251.2 | XP_011507553.1 | 13, 14 | 1121 | |||
XM_017000470.1 | XP_016855959.1 | 14, 15 | 1246 | |||
XM_017000471.1 | XP_016855960.1 | 13, 14 | 1108 | |||
XM_017000472.1 | XP_016855961.1 | 9, 10 | 732 | |||
GenBank mRNA | AK091207.1 | - | 12, 13 | 1012 | ||
BC022561.1 | - | 13, 14 | 1111 | 4793412 | ||
BC063877.1 | - | 13, 14 | 1090 | 30377843 |