Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 15 | 1465 | ||
NM_001300858.1 | NP_001287787.1 | 15 | 1616 | |||
NM_144977.4 | NP_659414.2 | 15 | 1465 | |||
XM_005244931.2 | XP_005244988.1 | 14 | 1405 | |||
XM_006711192.2 | XP_006711255.1 | 16 | 1375 | |||
XM_006711193.3 | XP_006711256.1 | 16 | 1507 | |||
XM_006711194.3 | XP_006711257.1 | 3 | 11503 | |||
XM_011509246.2 | XP_011507548.1 | 15 | 1326 | |||
XM_011509247.1 | XP_011507549.1 | 14 | 1266 | |||
XM_011509248.2 | XP_011507550.1 | 15 | 1259 | |||
XM_011509249.2 | XP_011507551.1 | 16 | 1564 | |||
XM_011509251.2 | XP_011507553.1 | 15 | 1328 | |||
XM_017000470.1 | XP_016855959.1 | 16 | 1453 | |||
XM_017000471.1 | XP_016855960.1 | 15 | 1315 | |||
XM_017000472.1 | XP_016855961.1 | 11 | 939 | |||
GenBank mRNA | BC022561.1 | - | 15 | 1318 | 4793412 | |
BC063877.1 | - | 15 | 1297 | 30377843 | ||
H19092.1 | - | 2 | 177 |