Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152889.2 | NP_690849.1 | 3 | 840 | ||
GenBank mRNA | AY120869.1 | - | 3 | 795 | ||
BC103896.1 | - | 4 | 614 | 40005101 | ||
BC103897.1 | - | 4 | 614 | 40005103 |