Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270391.1 | NP_001257320.1 | 3, 4 | 466 | ||
NM_177532.4 | NP_803876.1 | 3, 4 | 466 | |||
NM_201431.2 | NP_958834.1 | 3, 4 | 376 | |||
XM_011531687.2 | XP_011529989.1 | 3, 4 | 368 | |||
XM_011531688.1 | XP_011529990.1 | 4, 5 | 442 | |||
XM_011531689.2 | XP_011529991.1 | 3, 4 | 368 | |||
XM_011531690.2 | XP_011529992.1 | 2, 3 | 3002 | |||
XM_017007838.1 | XP_016863327.1 | 3, 4 | 368 | |||
GenBank mRNA | AK126346.1 | - | 3, 4 | 368 | ||
AY217664.1 | - | 3, 4 | 280 | |||
BC058835.1 | - | 3, 4 | 309 | 6668006 | ||
BX957171.1 | - | 3, 4 | 378 | |||
CR749283.1 | - | 3, 4 | 466 |