Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177455.3 | NP_803238.1 | 1 | 316 | ||
GenBank mRNA | BC113394.1 | - | 1 | 316 | 8322446 | |
BC113396.1 | - | 1 | 316 | 8322448 |