Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133497.3 | NP_598004.1 | 2 | 1723 | ||
GenBank mRNA | AF348983.1 | - | 2 | 1513 | ||
BC101352.1 | - | 2 | 1723 | 40025518 | ||
BC101353.2 | - | 2 | 1723 | 40025524 | ||
EL952823.1 | - | 1 | 71 |