Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003699.1 | 4 | 1511 | |||
GenBank mRNA | AB075859.1 | - | 1 | 457 | ||
AK098804.1 | - | 1 | 582 |