Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243168.1 | NP_001230097.1 | 2 | 427 | ||
NM_001243169.1 | NP_001230098.1 | 3 | 352 | |||
NM_138296.2 | NP_612153.2 | 2 | 427 | |||
GenBank mRNA | AF165312.1 | - | 3 | 271 | ||
AK130819.1 | - | 361 | ||||
AK310559.1 | - | 2 | 491 | |||
AK312223.1 | - | 2 | 360 | |||
BC069336.1 | - | 2 | 395 | 7262230 | ||
BC100771.1 | - | 2 | 369 | 40000277 | ||
BC100772.1 | - | 2 | 369 | 40000280 | ||
BC100773.2 | - | 2 | 369 | 40000282 | ||
BC100774.2 | - | 2 | 369 | 40000286 | ||
BC153829.1 | - | 2 | 369 | 40000274 | ||
U36759.1 | - | 2 | 427 | |||
U38996.1 | - | 2 | 312 |