Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030059.2 | NP_001025230.1 | 1, 2 | 396 | ||
NM_001318166.1 | NP_001305095.1 | 1, 2 | 396 | |||
NM_001318167.1 | NP_001305096.1 | 1, 2 | 396 | |||
NM_001318168.1 | NP_001305097.1 | 1, 2 | 396 | |||
NM_001318169.1 | NP_001305098.1 | 1, 2 | 396 | |||
NR_134516.1 | 1, 3 | 188 | ||||
XM_005269592.2 | XP_005269649.1 | 1, 2 | 396 | |||
XM_006717686.3 | XP_006717749.1 | 1, 2 | 396 | |||
XM_011539445.2 | XP_011537747.1 | 1, 2 | 396 | |||
XM_017015823.1 | XP_016871312.1 | 1, 2 | 396 | |||
XM_017015824.1 | XP_016871313.1 | 1, 2 | 396 | |||
GenBank mRNA | AK310178.1 | - | 1, 2 | 47 | ||
BC101267.2 | - | 2, 3 | 194 | |||
BC101268.2 | - | 2, 3 | 90 | 40024099 | ||
BC101269.1 | - | 1, 3 | 71 | |||
BC122535.1 | - | 2, 3 | 193 | |||
BC132787.1 | - | 1, 2 | 219 | 40146809 | ||
BI550027.1 | - | 1, 2 | 128 | |||
DB473853.2 | - | 1, 2 | 103 |