Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1DP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833768.1 | - | 1 | 2471 | ||
BC021729.1 | - | 4 | 457 | |||
BC125059.1 | - | 1 | 1520 | |||
BC125060.1 | - | 1 | 1517 |