Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152911.3 | NP_690875.1 | 3 | 836 | ||
NM_207127.2 | NP_997010.1 | 3 | 836 | |||
NM_207128.2 | NP_997011.1 | 3 | 836 | |||
NR_109763.1 | 3 | 836 | ||||
NR_109764.1 | 2 | 349 | ||||
NR_109765.1 | 2 | 349 | ||||
NR_109766.1 | 2 | 349 | ||||
GenBank mRNA | AF226657.1 | - | 2 | 576 | ||
AF312698.1 | - | 3 | 882 | |||
AK128001.1 | - | 3 | 720 | |||
AL834535.1 | - | 1 | 81 | |||
AY358418.1 | - | 3 | 776 | |||
AY541513.1 | - | 3 | 786 | |||
AY541514.1 | - | 2 | 299 | |||
AY541515.1 | - | 3 | 786 | |||
AY541516.1 | - | 3 | 812 | |||
AY541517.1 | - | 2 | 297 | |||
AY541518.1 | - | 2 | 299 | |||
AY541519.1 | - | 3 | 786 | |||
AY541520.1 | - | 1200 | ||||
AY541524.1 | - | 4 | 686 | |||
BC032778.1 | - | 3 | 847 | 4932036 |