Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152657.3 | NP_689870.3 | 3 | 652 | ||
XM_005258619.4 | XP_005258676.1 | 3 | 1014 | |||
XM_011526603.2 | XP_011524905.1 | 3 | 764 | |||
XM_017026451.1 | XP_016881940.1 | 2 | 858 | |||
GenBank mRNA | AF538036.1 | - | 3 | 657 | ||
AK057356.2 | - | 3 | 392 | |||
AK093032.1 | - | 2 | 600 | |||
BC035008.2 | - | 3 | 652 | 5269088 |