Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011547.2 | NP_001011547.2 | 9 | 1142 | ||
NM_001135181.1 | NP_001128653.1 | 10 | 1217 | |||
XM_011540924.2 | XP_011539226.1 | 9 | 2823 | |||
XM_011540925.2 | XP_011539227.1 | 9 | 2902 | |||
XM_011540926.2 | XP_011539228.1 | 8 | 1403 | |||
XM_011540927.1 | XP_011539229.1 | 7 | 1001 | |||
XM_011540928.1 | XP_011539230.1 | 9 | 1021 | |||
XM_011540929.2 | XP_011539231.1 | 9 | 2906 | |||
XM_017000558.1 | XP_016856047.1 | 9 | 2905 | |||
GenBank mRNA | AK131200.1 | - | 9 | 1095 | ||
AK296133.1 | - | 8 | 815 | |||
BC104857.1 | - | 9 | 1142 | 8143860 | ||
BC104863.1 | - | 9 | 1142 | 8143866 | ||
BC143570.1 | - | 10 | 1205 | 9052081 | ||
BC143572.1 | - | 10 | 1215 | 9052083 | ||
BC143587.1 | - | 8 | 936 | |||
BX648549.1 | - | 10 | 1171 |