Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTCAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173852.3 | NP_776251.1 | 1 | 53 | ||
GenBank mRNA | AK024167.1 | - | 1 | 29 | ||
AK311907.1 | - | 1 | 27 | |||
BC029806.1 | - | 1 | 40 | 5189880 | ||
BC057233.1 | - | 1 | 62 | 6463372 |