Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173545.2 | NP_775816.1 | 7 | 1238 | ||
GenBank mRNA | AK290841.1 | - | 7 | 1235 | ||
AK308186.1 | - | 6 | 1128 | |||
BC030711.2 | - | 3 | 434 | |||
BC041144.1 | - | 7 | 1187 | 6042653 |