Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144606.5 | NP_653207.1 | 4 | 521 | ||
NM_144997.5 | NP_659434.2 | 4 | 521 | |||
XM_011523714.2 | XP_011522016.1 | 4 | 518 | |||
XM_011523716.1 | XP_011522018.1 | 2 | 896 | |||
XM_011523718.2 | XP_011522020.1 | 3 | 402 | |||
XM_011523719.2 | XP_011522021.1 | 4 | 518 | |||
XM_011523721.2 | XP_011522023.1 | 3 | 267 | |||
XM_017024304.1 | XP_016879793.1 | 5 | 687 | |||
XM_017024305.1 | XP_016879794.1 | 5 | 702 | |||
XM_017024306.1 | XP_016879795.1 | 5 | 745 | |||
XM_017024307.1 | XP_016879796.1 | 5 | 707 | |||
XM_017024308.1 | XP_016879797.1 | 6 | 883 | |||
XM_017024309.1 | XP_016879798.1 | 4 | 518 | |||
GenBank mRNA | AF517523.1 | - | 4 | 472 | ||
AK126951.1 | - | 1 | 1358 | |||
AK127912.1 | - | 4 | 521 | |||
AK309336.1 | - | 4 | 518 | |||
BC015687.2 | - | 4 | 472 | 4684974 | ||
BQ423946.1 | - | 526 | ||||
CX759825.1 | - | 4 | 484 | |||
DA461022.1 | - | 4 | 518 | |||
DA944035.1 | - | 4 | 521 | |||
KU178888.1 | - | 1 | 17 | |||
KU178889.1 | - | 1 | 17 |