Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC202181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026921.1 | 6 | 2144 | |||
GenBank mRNA | AF086197.1 | - | 1 | 550 | ||
AK021623.1 | - | 1 | 527 |