Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3HCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136108.2 | NP_001129580.2 | 5 | 1084 | ||
NM_001301650.1 | NP_001288579.1 | 6 | 958 | |||
NR_125897.1 | 6 | 969 | ||||
GenBank mRNA | AK293465.1 | - | 5 | 1068 | ||
AL050297.1 | - | 5 | 1062 | |||
BC032421.1 | - | 5 | 1051 | |||
BC050572.1 | - | 4 | 1023 | |||
BC143822.1 | - | 6 | 1142 | 9052337 | ||
BC143827.1 | - | 6 | 1153 | |||
BC143829.1 | - | 5 | 1268 | 9052344 |