Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206559.1 | NP_001193488.1 | 3 | 702 | ||
NM_152705.2 | NP_689918.1 | 3 | 535 | |||
XM_005266414.2 | XP_005266471.1 | 4 | 1375 | |||
XM_017020622.1 | XP_016876111.1 | 2 | 402 | |||
XM_017020623.1 | XP_016876112.1 | 4 | 1295 | |||
GenBank mRNA | AF131855.1 | - | 1 | 1288 | ||
AK097973.1 | - | 4 | 622 | |||
AK307356.1 | - | 3 | 499 | |||
AK311833.1 | - | 3 | 422 | |||
BC006972.1 | - | 3 | 435 | |||
BC015319.2 | - | 3 | 444 | 3847651 | ||
BC018528.1 | - | 3 | 448 | 3865616 |