Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130159.2 | NP_001123631.1 | 4 | 882 | ||
NM_001130160.2 | NP_001123632.1 | 3 | 682 | |||
NM_001130161.2 | NP_001123633.1 | 4 | 3041 | |||
NM_152709.4 | NP_689922.3 | 4 | 3041 | |||
XM_011539454.2 | XP_011537756.1 | 4 | 2772 | |||
GenBank mRNA | AK057864.1 | - | 2 | 660 | ||
AK057891.1 | - | 2 | 1431 | |||
AK124681.1 | - | 4 | 2736 | |||
AK127878.1 | - | 2 | 2082 | |||
AY842014.1 | - | 4 | 2954 | |||
AY842015.1 | - | 4 | 795 | |||
AY842016.1 | - | 3 | 595 | |||
AY842017.1 | - | 4 | 2954 | |||
BC063627.1 | - | 2 | 1422 | |||
BC140011.1 | - | 4 | 882 |