Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM218 SIRNA Assays ›
Gene Symbol:
TMEM218
Gene Name:
transmembrane protein 218
Gene Aliases:
-
Chromosome Location:
Chr.11: 125094370 - 125111763 on Build GRCh38
UniGene ID:
Hs.731899
Species:
Human
siRNA ID
HSS153563
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM218
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080546.2
NP_001074015.1
5
763
NM_001258238.1
NP_001245167.1
5
641
NM_001258239.1
NP_001245168.1
5
738
NM_001258240.1
NP_001245169.1
7
801
NM_001258241.1
NP_001245170.1
6
742
NM_001258242.1
NP_001245171.1
5
729
NM_001258243.1
NP_001245172.1
6
832
NM_001258244.1
NP_001245173.1
5
632
NM_001258245.1
NP_001245174.1
7
794
NM_001258246.1
NP_001245175.1
5
714
NM_001258247.1
NP_001245176.1
5
617
NR_047586.1
5
594
NR_047587.1
3
377
NR_047588.1
5
682
NR_047589.1
4
446
NR_047590.1
5
619
NR_047591.1
5
505
NR_047592.1
5
585
XM_006718779.3
XP_006718842.1
7
942
XM_006718780.3
XP_006718843.1
6
839
XM_006718781.3
XP_006718844.1
7
896
XM_006718783.3
XP_006718846.1
5
705
XM_017017322.1
XP_016872811.1
6
729
XM_017017323.1
XP_016872812.1
5
655
XM_017017324.1
XP_016872813.1
5
657
GenBank mRNA
AI680968.1
-
1
71
AK091324.1
-
2
3940
AL833215.1
-
2
344
BC132716.1
-
5
600
40146738
BC132718.1
-
5
600
40146740
BC144270.1
-
6
703
9052790
BC144276.1
-
5
553
BC144277.1
-
5
634
9052797
BE513148.1
-
5
573
BE974033.1
-
3
222
BI192743.1
-
5
486
BI547955.1
-
5
391
BI829527.1
-
5
576
BI856825.1
-
5
507
BM042315.1
-
6
593
BQ223298.1
-
7
689
BQ225039.1
-
4
363
BQ420663.1
-
5
608
BQ653243.1
-
5
608
BQ681518.1
-
6
591
BU160644.1
-
5
438
BU598207.1
-
5
535
BU855096.1
-
5
529
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbzcs:80/10.239.166.68:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline