Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181514.1 | NP_852615.1 | 3, 4 | 253 | ||
NM_181515.1 | NP_852616.1 | 3, 4 | 266 | |||
XM_005273823.4 | XP_005273880.1 | 3, 4 | 250 | |||
GenBank mRNA | AK096756.1 | - | 3, 4 | 266 | ||
AK301989.1 | - | 3, 4 | 250 | |||
AV711404.1 | - | 3, 4 | 125 | |||
BC055088.1 | - | 3, 4 | 252 | 6457142 | ||
BM916618.1 | - | 3, 4 | 218 |