Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP32P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003554.1 | 4, 5 | 2578 | |||
GenBank mRNA | AJ012755.1 | - | 4, 5 | 2132 | ||
AK124785.1 | - | 9, 10 | 2498 | |||
AK308293.1 | - | 7, 8 | 1290 |