Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001971.2 | NP_001001971.1 | 10 | 1291 | ||
NM_001143773.1 | NP_001137245.1 | 13 | 1756 | |||
NM_001166698.1 | NP_001160170.1 | 13 | 1667 | |||
NM_198215.3 | NP_937858.2 | 12 | 1585 | |||
XM_005269618.4 | XP_005269675.2 | 12 | 1582 | |||
XM_005269619.4 | XP_005269676.2 | 10 | 1288 | |||
XM_006717702.3 | XP_006717765.1 | 14 | 1586 | |||
XM_006717703.3 | XP_006717766.1 | 14 | 1768 | |||
XM_011539490.2 | XP_011537792.1 | 13 | 1641 | |||
XM_011539491.2 | XP_011537793.1 | 14 | 1687 | |||
XM_017015885.1 | XP_016871374.1 | 13 | 1499 | |||
XM_017015886.1 | XP_016871375.1 | 13 | 1753 | |||
XM_017015887.1 | XP_016871376.1 | 13 | 1503 | |||
XM_017015888.1 | XP_016871377.1 | 14 | 1765 | |||
XM_017015889.1 | XP_016871378.1 | 14 | 1583 | |||
XM_017015890.1 | XP_016871379.1 | 14 | 1822 | |||
XM_017015891.1 | XP_016871380.1 | 11 | 1462 | |||
XM_017015892.1 | XP_016871381.1 | 11 | 1459 | |||
XM_017015893.1 | XP_016871382.1 | 13 | 1563 | |||
GenBank mRNA | AK289796.1 | - | 12 | 1511 | ||
AK294805.1 | - | 12 | 1500 | |||
AK299924.1 | - | 13 | 1756 | |||
AK316542.1 | - | 13 | 1663 | |||
BC036453.1 | - | 12 | 1543 | 5270033 | ||
BC064431.1 | - | 10 | 1291 | 5764753 |