Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002266.2 | NP_001002266.1 | 2 | 318 | ||
NM_001282866.1 | NP_001269795.1 | 2 | 318 | |||
NM_145021.5 | NP_659458.2 | 2 | 806 | |||
XM_005271804.2 | XP_005271861.1 | 2 | 807 | |||
XM_006717704.2 | XP_006717767.1 | 2 | 222 | |||
XM_011539492.2 | XP_011537794.1 | 3 | 858 | |||
XM_011539493.2 | XP_011537795.1 | 4 | 1099 | |||
XM_011539494.2 | XP_011537796.1 | 3 | 1048 | |||
XM_011539495.1 | XP_011537797.1 | 2 | 202 | |||
XM_017015894.1 | XP_016871383.1 | 3 | 858 | |||
GenBank mRNA | AA418494.1 | - | 1 | 19 | ||
AK307667.1 | - | 2 | 318 | |||
AK313340.1 | - | 2 | 275 | |||
BC025394.2 | - | 2 | 295 | 4830278 | ||
BC066988.1 | - | 2 | 807 | 5267214 | ||
BF195460.1 | - | 2 | 499 | |||
BG717635.1 | - | 2 | 300 | |||
BM906478.1 | - | 488 |