Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010870.2 | NP_001010870.1 | 1 | 5429 | ||
NM_001168359.1 | NP_001161831.1 | 1 | 5429 | |||
GenBank mRNA | AF039442.1 | - | 1 | 1366 | ||
AK125838.1 | - | 1 | 2517 | |||
BX648686.1 | - | 1 | 1961 | |||
CR936792.1 | - | 1 | 3780 | |||
EF185284.1 | - | 1 | 1914 |