Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181842.2 | NP_862825.1 | 2 | 413 | ||
XM_011514383.2 | XP_011512685.2 | 2 | 327 | |||
GenBank mRNA | BC043609.1 | - | 2 | 413 | 5767002 |
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178063.2 | NP_001171534.1 | 1 | 67 | ||
GenBank mRNA | AK298311.1 | - | 1 | 67 |