Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTURN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152793.2 | NP_690006.2 | 3 | 511 | ||
XM_005249652.3 | XP_005249709.1 | 3 | 454 | |||
GenBank mRNA | AK294982.1 | - | 3 | 241 | ||
AL834391.1 | - | 3 | 445 | |||
AY054121.1 | - | 3 | 307 | |||
BC062423.1 | - | 3 | 262 | |||
BI560774.1 | - | 3 | 515 | |||
DT217816.1 | - | 3 | 446 |