Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175887.2 | NP_787083.1 | 2 | 1466 | ||
XM_005249653.3 | XP_005249710.1 | 2 | 2269 | |||
XM_011515198.1 | XP_011513500.1 | 2 | 2264 | |||
XM_011515199.1 | XP_011513501.1 | 2 | 1178 | |||
XM_017011842.1 | XP_016867331.1 | 2 | 1476 | |||
GenBank mRNA | BC018144.1 | - | 2 | 1362 | ||
BC029131.1 | - | 2 | 1466 | 5189352 | ||
FJ857942.1 | - | 1 | 785 |