Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM185A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145268.1 | NP_001138740.1 | 1 | 644 | ||
XM_006715896.3 | XP_006715959.1 | 1 | 650 | |||
XM_011515922.2 | XP_011514224.1 | 1 | 651 | |||
XM_011515923.2 | XP_011514225.1 | 1 | 633 | |||
XM_011515924.2 | XP_011514226.1 | 1 | 651 | |||
XM_011515925.2 | XP_011514227.1 | 1 | 651 | |||
XM_011515926.2 | XP_011514228.1 | 1 | 651 | |||
XM_017011846.1 | XP_016867335.1 | 1 | 651 | |||
XM_017011847.1 | XP_016867336.1 | 1 | 651 | |||
XM_017011848.1 | XP_016867337.1 | 1 | 651 | |||
XM_017011849.1 | XP_016867338.1 | 1 | 633 | |||
GenBank mRNA | BC029175.1 | - | 1 | 625 | ||
DR006842.1 | - | 1 | 564 |