Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139021.2 | NP_620590.2 | 3, 4 | 227 | ||
XM_011516925.2 | XP_011515227.1 | 3, 4 | 1065 | |||
XM_011516926.2 | XP_011515228.1 | 3, 4 | 316 | |||
XM_011516927.2 | XP_011515229.1 | 3, 4 | 1008 | |||
XM_011516928.2 | XP_011515230.1 | 3, 4 | 1065 | |||
XM_017013218.1 | XP_016868707.1 | 3, 4 | 1065 | |||
GenBank mRNA | AY065978.1 | - | 3, 4 | 219 | ||
AY994058.1 | - | 3, 4 | 186 | |||
BC028034.1 | - | 3, 4 | 224 | 5242481 | ||
BP357805.1 | - | 3, 4 | 227 |