Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002257.2 | NP_001002257.1 | 6 | 1128 | ||
NM_001304445.1 | NP_001291374.1 | 8 | 1533 | |||
NM_001304446.1 | NP_001291375.1 | 6 | 1137 | |||
NM_182551.4 | NP_872357.2 | 7 | 1320 | |||
XM_005264244.1 | XP_005264301.1 | 7 | 1282 | |||
XM_005264245.3 | XP_005264302.1 | 6 | 1134 | |||
XM_011532741.2 | XP_011531043.1 | 7 | 1271 | |||
XM_011532742.2 | XP_011531044.1 | 7 | 1098 | |||
XM_011532743.2 | XP_011531045.1 | 6 | 1085 | |||
XM_011532744.2 | XP_011531046.1 | 7 | 1280 | |||
XM_017003746.1 | XP_016859235.1 | 7 | 1165 | |||
GenBank mRNA | AK296965.1 | - | 6 | 1105 | ||
AK297262.1 | - | 6 | 1117 | |||
AK302562.1 | - | 8 | 1486 | |||
AK311294.1 | - | 1, 2 | 106 | |||
AY358702.1 | - | 7 | 1289 | |||
BC146817.1 | - | 6 | 1118 | 8860419 | ||
BX647953.1 | - | 6 | 1085 |