Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099409.1 | NP_001092879.1 | 10 | 3487 | ||
XM_005273867.3 | XP_005273924.1 | 12 | 1900 | |||
XM_005273868.3 | XP_005273925.1 | 11 | 1725 | |||
XM_005273869.3 | XP_005273926.1 | 11 | 1427 | |||
XM_005273870.3 | XP_005273927.1 | 10 | 1252 | |||
XM_006718486.3 | XP_006718549.1 | 11 | 3524 | |||
XM_011544886.2 | XP_011543188.1 | 3 | 284 | |||
XM_017017467.1 | XP_016872956.1 | 11 | 3274 | |||
XM_017017468.1 | XP_016872957.1 | 11 | 3251 | |||
XM_017017469.1 | XP_016872958.1 | 9 | 1215 | |||
GenBank mRNA | AK024451.1 | - | 6 | 2899 | ||
AL713799.1 | - | 1 | 2846 | |||
AL834433.1 | - | 10 | 3184 |