Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143917.1 | NP_001137389.1 | 2 | 488 | ||
NM_153374.2 | NP_699205.1 | 2 | 1079 | |||
XM_017022057.1 | XP_016877546.1 | 2 | 471 | |||
GenBank mRNA | AK129759.1 | - | 2 | 558 | ||
AY125955.1 | - | 2 | 1068 | |||
BC022075.1 | - | 2 | 624 | 4799858 | ||
BC033515.1 | - | 2 | 1076 | 5175226 | ||
BQ446302.1 | - | 2 | 627 | |||
CR936638.1 | - | 1 | 118 |