Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013625.3 | NP_001013647.2 | 1, 2 | 30 | ||
GenBank mRNA | DT215043.1 | - | 1, 2 | 30 |