Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL4I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258017.1 | NP_001244946.1 | 5 | 795 | ||
NM_001258018.1 | NP_001244947.1 | 5 | 877 | |||
NM_152899.1 | NP_690863.1 | 3 | 268 | |||
NM_172374.2 | NP_758962.1 | 5 | 819 | |||
NR_047577.1 | 6 | 854 | ||||
GenBank mRNA | AF293462.1 | - | 3 | 268 | ||
AJ880386.1 | - | 5 | 694 | |||
AJ880387.1 | - | 5 | 671 | |||
AJ880388.1 | - | 5 | 733 | |||
AJ880389.1 | - | 6 | 731 | |||
AY358933.1 | - | 3 | 268 | |||
BC026103.1 | - | 5 | 723 | |||
BC064378.1 | - | 3 | 254 | 5725335 | ||
BC090852.1 | - | 3 | 255 | 6191892 | ||
BC131625.1 | - | 5 | 708 | 40117891 | ||
DQ079587.1 | - | 5 | 819 | |||
DQ079588.1 | - | 5 | 877 | |||
DQ079589.1 | - | 5 | 795 |