Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098483.2 | NP_001091953.1 | 4 | 305 | ||
NM_001301827.1 | NP_001288756.1 | 3 | 236 | |||
NM_001301828.1 | NP_001288757.1 | 4 | 512 | |||
NM_198472.2 | NP_940874.2 | 4 | 305 | |||
XM_011539670.2 | XP_011537972.1 | 4 | 311 | |||
XM_011539671.2 | XP_011537973.1 | 4 | 311 | |||
XM_011539672.2 | XP_011537974.1 | 4 | 311 | |||
XM_011539673.2 | XP_011537975.1 | 4 | 311 | |||
XM_011539674.2 | XP_011537976.1 | 4 | 311 | |||
XM_011539675.2 | XP_011537977.1 | 4 | 311 | |||
XM_017016094.1 | XP_016871583.1 | 4 | 509 | |||
GenBank mRNA | AK129527.1 | - | 4 | 305 | ||
AK310070.1 | - | 3 | 713 | |||
BC129818.1 | - | 4 | 305 | |||
BC129819.1 | - | 4 | 305 | 40113019 | ||
BM009909.1 | - | 3 | 168 | |||
BU849497.1 | - | 4 | 488 | |||
BX119427.1 | - | 4 | 231 | |||
HY163253.1 | - | 4 | 300 |