Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134363.2 | NP_001127835.2 | 2 | 384 | ||
XM_011539697.2 | XP_011537999.1 | 2 | 222 | |||
XM_017016103.1 | XP_016871592.1 | 2 | 1642 | |||
XM_017016104.1 | XP_016871593.1 | 2 | 3858 | |||
GenBank mRNA | EU822950.1 | - | 2 | 384 |