Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUXAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC066547.1 | - | 1 | 1657 | 5481815 |
Gene Symbol: LINC01296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096951.1 | - | 1 | 2181 |
Gene Symbol: LOC283028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136644.1 | 1 | 1174 | |||
GenBank mRNA | AK092783.1 | - | 1 | 1174 |