Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080407.2 | NP_001073876.2 | 20 | 2877 | ||
XM_011542566.2 | XP_011540868.1 | 19 | 2947 | |||
XM_011542568.2 | XP_011540870.1 | 19 | 2911 | |||
XM_011542569.2 | XP_011540871.1 | 15 | 2536 | |||
XM_011542570.2 | XP_011540872.1 | 15 | 2500 | |||
XM_011542572.1 | XP_011540874.1 | 15 | 2095 | |||
XM_011542573.1 | XP_011540875.1 | 13 | 2031 | |||
XM_011542574.2 | XP_011540876.1 | 13 | 2154 | |||
XM_017017155.1 | XP_016872644.1 | 20 | 3775 | |||
XM_017017156.1 | XP_016872645.1 | 16 | 3400 | |||
GenBank mRNA | AK074712.1 | - | 11 | 1584 | ||
AK124514.1 | - | 6 | 3796 |