Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176818.2 | NP_789788.1 | 2, 3 | 291 | ||
GenBank mRNA | AK094319.1 | - | 2, 3 | 285 | ||
BC064523.1 | - | 2, 3 | 262 | |||
BC107145.1 | - | 2, 3 | 248 | 40031296 | ||
BC107146.1 | - | 2, 3 | 241 |