Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7E156P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002171.2 | 2 | 1246 | |||
GenBank mRNA | AK097469.1 | - | 2 | 1247 | ||
AK097490.1 | - | 2 | 485 | |||
AK097573.1 | - | 2 | 524 |