Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207117.2 | NP_997000.2 | 6 | 857 | ||
GenBank mRNA | AB073385.1 | - | 7 | 781 | ||
AK092001.1 | - | 6 | 861 | |||
AY569438.1 | - | 6 | 857 | |||
BC137253.1 | - | 6 | 837 | 9021250 | ||
BC137254.1 | - | 6 | 837 | 9021251 | ||
BC144549.1 | - | 6 | 898 | |||
BX248260.1 | - | 6 | 846 |