Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAEP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024178.1 | 7 | 1075 | |||
GenBank mRNA | AK094896.1 | - | 7 | 1075 | ||
BC132826.1 | - | 5 | 934 | |||
BC144329.1 | - | 5 | 934 | 9052849 |