Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046174.2 | 6 | 840 | |||
NR_046454.1 | 5 | 620 | ||||
NR_046455.1 | 4 | 457 | ||||
NR_046456.1 | 7 | 931 | ||||
NR_046457.1 | 6 | 711 | ||||
GenBank mRNA | AB593072.1 | - | 7 | 931 | ||
AB593073.1 | - | 4 | 457 | |||
AB593075.1 | - | 6 | 709 | |||
BC021928.1 | - | 6 | 805 | |||
BC040725.2 | - | 5 | 599 |