Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019121.1 | NP_061994.1 | 10, 11 | 1454 | ||
XM_006723159.2 | XP_006723222.1 | 10, 11 | 1454 |