Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012753.1 | NP_001012771.1 | 4 | 1132 | ||
GenBank mRNA | AK092240.1 | - | 4 | 1132 | ||
AK299182.1 | - | 4 | 1164 | |||
AK299224.1 | - | 2 | 935 | |||
BC105711.1 | - | 4 | 1095 | 40024621 | ||
BC105712.1 | - | 4 | 1095 | 40024618 | ||
BC105713.1 | - | 4 | 1095 | 40024617 |