Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM150B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085488.2 | NP_001078957.1 | 6 | 444 | ||
NM_001282011.1 | NP_001268940.1 | 6 | 493 | |||
NR_104066.1 | 5 | 425 | ||||
NR_104071.1 | 5 | 376 | ||||
XM_005258812.3 | XP_005258869.1 | 8 | 773 | |||
XM_011526850.2 | XP_011525152.1 | 7 | 652 | |||
XM_011526851.2 | XP_011525153.1 | 7 | 650 | |||
XM_011526852.2 | XP_011525154.1 | 7 | 650 | |||
XM_011526853.2 | XP_011525155.1 | 6 | 564 | |||
XM_011526854.2 | XP_011525156.1 | 5 | 729 | |||
XM_011526855.2 | XP_011525157.1 | 7 | 602 | |||
XM_011526856.2 | XP_011525158.1 | 5 | 681 | |||
XM_011526857.2 | XP_011525159.1 | 4 | 344 | |||
XM_017026667.1 | XP_016882156.1 | 7 | 664 | |||
XM_017026668.1 | XP_016882157.1 | 4 | 367 | |||
GenBank mRNA | AU311823.1 | - | 6 | 444 | ||
BC171893.1 | - | 5 | 341 | |||
BC171901.1 | - | 5 | 390 | |||
BC171904.1 | - | 6 | 454 | 9054558 |